Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

703052

Sigma-Aldrich

Dicyclohexylboron trifluoromethanesulfonate

97%

Synonym(s):

Dicyclohexylboron triflate, Dicyclohexylboryl trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22BF3O3S
CAS Number:
Molecular Weight:
326.18
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

59-65 °C

functional group

fluoro
triflate

storage temp.

2-8°C

SMILES string

FC(F)(F)S(=O)(=O)OB(C1CCCCC1)C2CCCCC2

InChI

1S/C13H22BF3O3S/c15-13(16,17)21(18,19)20-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

FBVDLYBQGQLANQ-UHFFFAOYSA-N

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service