Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

406139

Sigma-Aldrich

Chlorodicyclohexylborane

98%

Synonym(s):

Dicyclohexylboron chloride, Dicyclohexylboryl chloride, Dicyclohexylchloroborane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2BCl
CAS Number:
Molecular Weight:
212.57
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

Quality Level

Assay

98%

form

liquid

reaction suitability

reagent type: reductant

bp

100-101 °C/1 mmHg (lit.)

density

0.97 g/mL at 25 °C (lit.)

SMILES string

ClB(C1CCCCC1)C2CCCCC2

InChI

1S/C12H22BCl/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

PBBOKJIYEZCTEH-UHFFFAOYSA-N

Related Categories

Application

A convenient reagent for regio- and stereoselective enolboration of ketones and other carbonyl derivatives.
Reactant for:
  • Aldol addition reactions
  • Mukaiyama aldol addition
  • Microwave assisted ring closing metathesis reactions
  • C-alkylation of aromatic aldimines
  • Reversible ketone-ketone aldol reactions
  • Syn- and anti-stereoselective aldol reaction hexenone with aldehydes

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 57, 499-499 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service