Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

411124

Sigma-Aldrich

Chlorodicyclohexylborane solution

1 M in hexanes

Synonym(s):

Dicyclohexylboron chloride, Dicyclohexylboryl chloride, Dicyclohexylchloroborane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2BCl
CAS Number:
Molecular Weight:
212.57
Beilstein:
3233942
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1 M in hexanes

bp

68-69 °C

density

0.752 g/mL at 25 °C

SMILES string

ClB(C1CCCCC1)C2CCCCC2

InChI

1S/C12H22BCl/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

PBBOKJIYEZCTEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Aldol addition reactions
  • Mukaiyama aldol addition
  • Microwave assisted ring closing metathesis reactions
  • C-alkylation of aromatic aldimines
  • Reversible ketone-ketone aldol reactions
  • Syn- and anti-stereoselective aldol reaction hexenone with aldehydes

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service