658693
Pd(II) EnCat® BINAP30
extent of labeling: 0.39 mmol/g Pd loading, BINAP 0.25, Pd 1.0
Synonym(s):
Palladium acetate and BINAP, microencapsulated in polyurea matrix
About This Item
Recommended Products
composition
BINAP, 0.25
Pd, 1.0
reaction suitability
core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
extent of labeling
0.39 mmol/g Pd loading
General description
Application
Features and Benefits
- Low residual metal levels in crude product.
- Easy recovery of catalyst.
- Compatibility of immobilized metal with various ligands to effect C–C bond forming reactions.
- Safer and easier to handle than Pd/C for hydrogenation.
- Efficiency and economy gains through recovery and recycling.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Sol. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation
Target Organs
Nervous system
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service