Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

644722

Sigma-Aldrich

Pd EnCat® 40

extent of labeling: 0.4 mmol/g Pd loading

Synonym(s):

Palladium acetate microencapsulated in polyurea matrix

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

contains

0.37-0.44 mmol/g palladium

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

extent of labeling

0.4 mmol/g Pd loading

General description

It is an encapsulated catalyst (EnCat®), wherein palladium acetate is microencapsulated within a polyurea matrix, which prevents palladium leaching.

Application

Recyclable, immobilized catalyst system that simplifies removal of Pd in C-C bond forming and reduction processes.

Features and Benefits

The advantages of using Pd EnCat® catalysts are:
  • low residual metal levels in crude product
  • easy recovery of catalyst
  • compatibility of immobilized metal with various ligands to effect C-C bond forming reactions
  • safer and easier to handle than Pd/C for hydrogenation
  • efficiency and economy gains through recovery and recycling

Other Notes

Matrix content 40%

Legal Information

PdEncat® technology.
EnCat is a registered trademark of Reaxa Ltd.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Sol. 2 - Repr. 1B - Skin Irrit. 2

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven V Ley et al.
Chemical communications (Cambridge, England), (10), 1134-1135 (2002-07-19)
Palladium(II) acetate microencapsulated in polyurea (MC-[Pd]) is an economical and versatile heterogeneous catalyst for a range of phosphine-free cross-coupling reactions in both conventional solvents and supercritical carbon dioxide (scCO2); the catalyst can be recovered by a simple filtration and recycled
Steven V Ley et al.
Organic letters, 5(24), 4665-4668 (2003-11-25)
[reaction: see text] Pd nanoparticles (approximately 2 nm in size) microencapsulated in polyurea is an efficient and recyclable catalyst for reductive ring-opening hydrogenolysis of epoxides, using either HCOOH/Et(3)N or H(2) as a hydrogen donor.
Yu, J.Q., et al.
Chemical Communications (Cambridge, England), 678-697 (2003)

Related Content

Homogeneous palladium catalysts are widely utilized due to their versatility, reactivity and functional group tolerance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service