Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

590231

Sigma-Aldrich

Di(acetato)dicyclohexylphenylphosphinepalladium(II), polymer-bound FibreCat®

Pd ~5 %, extent of labeling: 0.4-0.6 mmol/g PPH3 ligand content loading

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

product line

FibreCat®

Quality Level

form

solid

composition

Pd, ~5%

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reaction type: solution phase peptide synthesis
reagent type: catalyst
reaction type: Coupling Reactions

extent of labeling

0.4-0.6 mmol/g PPH3 ligand content loading

SMILES string

[Pd+2].P(C3CCCCC3)(C2CCCCC2)c1ccccc1.[O-]C(=O)C.[O-]C(=O)C

InChI key

ZMMYBYFXUHUTSH-UHFFFAOYSA-L

Application

Supported catalyst used in Suzuki coupling of less reactive aryl chloride with boronic acids. This supported catalyst is generally more selective than the free homogeneous catalyst, and palladium leaching is very low (<2 ppm).
The resin can also be recycled.

Legal Information

FibreCat is a registered trademark of Johnson Matthey PLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colacot, T.J.
Organometallics, 21, 3301-3301 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service