Skip to Content
MilliporeSigma
All Photos(1)

Documents

655392

Sigma-Aldrich

Bis-tris-d19

98 atom % D, 98% (CP)

Synonym(s):

2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol-d19

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(DOCD2CD2)2NC(CD2OD)3
CAS Number:
Molecular Weight:
228.36
MDL number:
UNSPSC Code:
12161705
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

solid

technique(s)

NMR: suitable
bio NMR: suitable

mp

104 °C (lit.)

mass shift

M+19

SMILES string

[2H]OC([2H])([2H])C([2H])([2H])N(C([2H])([2H])C([2H])([2H])O[2H])C(C([2H])([2H])O[2H])(C([2H])([2H])O[2H])C([2H])([2H])O[2H]

InChI

1S/C8H19NO5/c10-3-1-9(2-4-11)8(5-12,6-13)7-14/h10-14H,1-7H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,10D,11D,12D,13D,14D

InChI key

OWMVSZAMULFTJU-ZMHMTVDGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Friedemann Flügge et al.
Journal of biomolecular NMR, 70(4), 245-259 (2018-04-28)
Human blood group A and B glycosyltransferases (GTA, GTB) are highly homologous glycosyltransferases. A number of high-resolution crystal structures is available showing that these enzymes convert from an open conformation into a catalytically active closed conformation upon substrate binding. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service