Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

151823

Sigma-Aldrich

Chloroform-d

99.8 atom % D

Synonym(s):

Deuterochloroform

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDCl3
CAS Number:
Molecular Weight:
120.38
Beilstein:
1697633
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.8 atom % D

Quality Level

Assay

≥99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.444 (lit.)

bp

60.9 °C (lit.)

mp

−64 °C (lit.)

density

1.500 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]C(Cl)(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform-d (Deuterochloroform, CDCl3), deuterated chloroform, is a standard purity solvent for NMR (Nuclear Magnetic Resonance) analyses. It is widely employed in high resolution NMR studies due to its high chemical and isotopic purity. Quantitaive infrared spectral investigations of carbon-deuterium stretching bands of chloroform-d in various organic solvents have been reported. Raman difference spectroscopic studies of mixtures of chloroform-d and liquid chloroform have been conducted to evaluate frequency shifts in the ν1 and ν2 bands of CHCl3 and CDCl3.

Application

Chloroform-d has been employed as solvent in a 1H NMR (Proton Nuclear Magnetic Resonance) spectroscopic study. It has been employed as solvent during the 31PNMR (Phosphorus-31 nuclear magnetic resonance) spectral investigation of triphenyl phosphate and tris(2-chloroethyl) phosphate. It has also been used as the deuterated solvent in the 1H NMR spectral studies of 2-(4-bromobenzyl)-3-hydroxypropanoic acid and 3-hydroxy-2-(4-methoxybenzyl)propanoic acid. It has also been used as the deuterated solvent for the 1HNMR spectral studies of 3,3′-dihydroxy-4,4′-diamino-biphenyl (HAB) and 2,2′-bis-(3,4-dicarboxy-phenyl) hexafluoropropane dianhydride (6FDA) polyimide.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas sorption and characterization of thermally rearranged polyimides based on 3, 3'-dihydroxy-4, 4'-diamino-biphenyl (HAB) and 2, 2'-bis-(3, 4-dicarboxyphenyl) hexafluoropropane dianhydride (6FDA)
Smith.PZ, et al.
Journal of Membrane Science, 415, 558-567 (2012)
Hyunwoo Joo et al.
Science advances, 7(1) (2021-02-02)
Personalized biomedical devices have enormous potential to solve clinical challenges in urgent medical situations. Despite this potential, a device for in situ treatment of fatal seizures using pharmaceutical methods has not been developed yet. Here, we present a novel treatment
Method development in quantitative NMR towards metrologically traceable organic certified reference materials used as 31P qNMR standards.
Weber M, et al.
Analytical and Bioanalytical Chemistry, 407(11), 3115-3123 (2015)
Selective synthesis of 3-hydroxy acids from Meldrum's acids using SmI2-H2O
Szostak M, et al.
Nature Protocols, 7(5), 970-977 (2012)
Michal Szostak et al.
Nature protocols, 7(5), 970-977 (2012-04-28)
The single-step synthesis of 3-hydroxy carboxylic acids from readily available Meldrum's acids involves a selective monoreduction using a SmI(2)-H(2)O complex to give products in high crude purity, and it represents a considerable advancement over other methods for the synthesis of

Articles

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

See All

Related Content

Widest range of NMR solvents with excellent chemical purity and the highest isotopic enrichment possible.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service