Skip to Content
MilliporeSigma
All Photos(1)

Documents

BI0345

Sigma-Aldrich

Capping Reagent B 16% n-Methylimidazole in THF

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006

vapor pressure

173 hPa ( 20 °C)

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

bp

65-66 °C/1013 hPa (Information refers to the main component.)

mp

-108.5 °C (Information refers to the main component.)

transition temp

flash point -22 °C (Information refers to the main component.)

shipped in

ambient

storage temp.

2-30°C

Application

BI0345-0450 is packaged in 28/405 glass bottles
BI0345-0950 is packaged in 38/430 glass bottles
BI0345-0960 is packaged in 28/405 glass bottles
BI0345-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation

Analysis Note

Assay (1-methylimidazole): 15.6-16.4% v/v
Assay (tetrahydrofuran): 83.6-84.4% v/v
Color (APHA): 10 max
Form: Clear liquid
Water (H2O): 300ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-7.6 °F - closed cup - Information refers to the main ingredient.

Flash Point(C)

-22 °C - closed cup - Information refers to the main ingredient.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service