Skip to Content
MilliporeSigma
All Photos(1)

Documents

492612

Sigma-Aldrich

Ethylene oxide-13C2

≥99 atom % 13C, ≥99% (CP), contains hydroquinone as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2H4O
CAS Number:
Molecular Weight:
46.04
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

contains

hydroquinone as stabilizer

refractive index

n20/D 1.3597 (lit.)

bp

10.7 °C (lit.)

mp

−111 °C (lit.)

density

0.859 g/mL at 25 °C

application(s)

environmental

mass shift

M+2

SMILES string

[13CH2]1[13CH2]O1

InChI

1S/C2H4O/c1-2-3-1/h1-2H2/i1+1,2+1

InChI key

IAYPIBMASNFSPL-ZDOIIHCHSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard 500 mg quantity is packaged in a Sure/Pac cylinder with stainless steel 1/4" male NPT valve. Nominal gas pressure at 21ºC is 7 psig for 500 mg. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel hose adapter Z146838 or stainless steel body mini gas regulator Z513547 is recommended.

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Chem. Unst. Gas A - Eye Dam. 1 - Flam. Gas 1A - Muta. 1B - Press. Gas Liquefied gas - Repr. 1B - Skin Corr. 1B - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Nervous system, Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service