Skip to Content
MilliporeSigma
All Photos(1)

Documents

570052

Sigma-Aldrich

Ethylene carbonate-13C3

99 atom % 13C, 97% (CP)

Synonym(s):

1,3-Dioxolan-2-one-2,4,5-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3H4O3
CAS Number:
Molecular Weight:
91.04
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

solid

mass shift

M+3

SMILES string

O=[13C]1O[13CH2][13CH2]O1

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2/i1+1,2+1,3+1

InChI key

KMTRUDSVKNLOMY-VMIGTVKRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service