Skip to Content
MilliporeSigma

489352

Ethylene-13C2

99 atom % 13C

Synonym(s):

Ethene-13C2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H213C=13CH2
CAS Number:
Molecular Weight:
30.04
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12352119
MDL number:
Isotopic purity:
99 atom % 13C
Assay:
99% (CP)
Mass shift:
M+2
Form:
gas
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

0.97 (vs air)

Quality Segment

isotopic purity

99 atom % 13C

assay

99% (CP)

form

gas

autoignition temp.

842 °F

bp

−104 °C (lit.)

mp

−169 °C (lit.)

mass shift

M+2

SMILES string

[13CH2]=[13CH2]

InChI

1S/C2H4/c1-2/h1-2H2/i1+1,2+1

InChI key

VGGSQFUCUMXWEO-ZDOIIHCHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.

Other Notes

Stainless steel control valve Z146951 or Z187224 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986


Signal Word

Danger

Hazard Codes

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas - STOT SE 3

Target Organs

Central nervous system

Storage Class

2A - Gases

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library