Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

457833

Sigma-Aldrich

Ethylene-d4 oxide

≥98 atom % D, ≥99% (CP), contains hydroquinone as stabilizer

Synonym(s):

Deuterated ethylene oxide, Oxirane-D4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2D4O
CAS Number:
Molecular Weight:
48.08
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12

vapor density

1.52 (vs air)

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

contains

hydroquinone as stabilizer

technique(s)

protein expression: suitable

bp

10.7 °C (lit.)

mp

-111 °C (lit.)

density

0.896 g/mL at 25 °C

application(s)

environmental

mass shift

M+4

SMILES string

[2H]C1([2H])OC1([2H])[2H]

InChI

1S/C2H4O/c1-2-3-1/h1-2H2/i1D2,2D2

InChI key

IAYPIBMASNFSPL-LNLMKGTHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

Shipping of this product by air freight is forbidden.
Outside of the U.S., ocean freight shipment is required and will result in substantial additional expense.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z148504 and Z148512 and a "T"-purge valve Z151661 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Chem. Unst. Gas A - Eye Dam. 1 - Flam. Gas 1A - Muta. 1B - Press. Gas Liquefied gas - Repr. 1B - Skin Corr. 1B - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Nervous system, Respiratory system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service