Skip to Content
MilliporeSigma
All Photos(1)

Documents

491667

Sigma-Aldrich

Acetone-13C3

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313CO13CH3
CAS Number:
Molecular Weight:
61.06
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.359 (lit.)

bp

56 °C (lit.)

mp

−94 °C (lit.)

density

0.832 g/mL at 25 °C

mass shift

M+3

SMILES string

[13CH3][13C]([13CH3])=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3/i1+1,2+1,3+1

InChI key

CSCPPACGZOOCGX-VMIGTVKRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service