Skip to Content
MilliporeSigma
All Photos(1)

Documents

277177

Sigma-Aldrich

Methanol-13C

99 atom % 13C

Synonym(s):

13C Labeled methanol, Methyl alcohol-13C, Methyl-13C alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3OH
CAS Number:
Molecular Weight:
33.03
Beilstein:
1697016
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable
bio NMR: suitable

refractive index

n20/D 1.329 (lit.)

bp

65.4 °C (lit.)

mp

-98 °C (lit.)

density

0.815 g/mL at 25 °C

format

neat

mass shift

M+1

SMILES string

[13CH3]O

InChI

1S/CH4O/c1-2/h2H,1H3/i1+1

InChI key

OKKJLVBELUTLKV-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

13CH3OH is isotopically enriched form of methyl alcohol.

Application

Methyl-13C alcohol is an external reference for the assignment of polymer constitution using nuclear magnetic resonance spectroscopy. It is used to study the proton transfer processes, which offering possibilities for the quantitative mechanism studies of proton-related catalytic reactions. This isotope-enrich solvent is used to confirm the distribution and arrangement of metals from metal organic frame work (MOF) using host-guest interaction study. Also used as trideuteromethylation reagent for the synthesis of deuterated drugs.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Lu Wang et al.
Nature communications, 7, 11918-11918 (2016-06-18)
Proton transfer (PT) processes in solid-liquid phases play central roles throughout chemistry, biology and materials science. Identification of PT routes deep into the realistic catalytic process is experimentally challenging, thus leaving a gap in our understanding. Here we demonstrate an
Sven Bodenstedt et al.
Nature communications, 12(1), 4041-4041 (2021-07-02)
Optically pumped magnetometers (OPMs) based on alkali-atom vapors are ultra-sensitive devices for dc and low-frequency ac magnetic measurements. Here, in combination with fast-field-cycling hardware and high-resolution spectroscopic detection, we demonstrate applicability of OPMs in quantifying nuclear magnetic relaxation phenomena. Relaxation
Monica I Espinosa et al.
Nature communications, 11(1), 5564-5564 (2020-11-06)
Utilising one-carbon substrates such as carbon dioxide, methane, and methanol is vital to address the current climate crisis. Methylotrophic metabolism enables growth and energy generation from methanol, providing an alternative to sugar fermentation. Saccharomyces cerevisiae is an important industrial microorganism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service