Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

490474

Sigma-Aldrich

Oxygen-18O2

97 atom % 18O

Synonym(s):

18O Labeled oxygen

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
18O2
CAS Number:
Molecular Weight:
36.00
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

vapor density

1.11 (vs air)

isotopic purity

97 atom % 18O

Assay

99% (CP)

bp

−183 °C (lit.)

mp

−218 °C (lit.)

mass shift

M+4

SMILES string

[18O]=[18O]

InChI

1S/O2/c1-2/i1+2,2+2

InChI key

MYMOFIZGZYHOMD-XPULMUKRSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard 500 mL quantitiy is packaged in a 25 mL Sure/Pac cylinder with brass 1/4" male NPT valve. Nominal gas pressure at 21ºC is 220 psig for 500 mL.

Standard 1 L quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180. Nominal gas pressure at 21ºC is 20 psig for 1 L.

Because of the relatively low cylinder pressures, care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended for 500 mL Sure/Pac cylinder (U.S only). For 1 L cylinder, either stainless steel control valve Z146951 or Z187224 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

Flame over circleGas cylinder

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Gas 1 - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maxwell A Ware et al.
Plant physiology, 183(4), 1735-1748 (2020-05-28)
The green alga Desmodesmus armatus is an emerging biofuel platform that produces high amounts of lipids and biomass in mass culture. We observed D. armatus in light-limiting, excess-light, and sinusoidal-light environments to investigate its photoacclimation behaviors and the mechanisms by
Sui So et al.
Physical chemistry chemical physics : PCCP, 16(45), 24954-24964 (2014-10-18)
β-Hydroxyperoxyl radicals are formed during atmospheric oxidation of unsaturated volatile organic compounds such as isoprene. They are intermediates in the combustion of alcohols. In these environments the unimolecular isomerization and decomposition of β-hydroxyperoxyl radicals may be of importance, either through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service