Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

902187

Sigma-Aldrich

Water-18O

(for PET), ≥98 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H218O
CAS Number:
Molecular Weight:
20.02
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.12

description

Normalized with respect to hydrogen

Quality Level

isotopic purity

≥98 atom % 18O

Assay

≥99%

form

liquid

technique(s)

PET imaging: suitable

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.11 g/mL at 20 °C (lit.)

compatibility

(for PET)

mass shift

M+2

storage temp.

room temp

SMILES string

[18OH2]

InChI

1S/H2O/h1H2/i1+2

InChI key

XLYOFNOQVPJJNP-NJFSPNSNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Appropriate for PET applications.

Packaging

10 g, 20 g, and 50 g pack sizes are packaged in a GMP environment and are in septum sealed bottles.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service