Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

593141

Sigma-Aldrich

Oxygen-18O2/Nitrogen(RG) Gas Mixture

ratio (1:4), 99 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12142200

isotopic purity

99 atom % 18O

form

gas

composition

ratio (1:4)

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Ox. Gas 1 - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service