Skip to Content
MilliporeSigma
All Photos(1)

Documents

486868

Sigma-Aldrich

Methane-d3

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHD3
CAS Number:
Molecular Weight:
19.06
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

vapor density

0.55 (vs air)

Quality Level

isotopic purity

98 atom % D

Assay

99% (CP)

bp

−161 °C (lit.)

mp

−183 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])[2H]

InChI

1S/CH4/h1H4/i1D3

InChI key

VNWKTOKETHGBQD-FIBGUPNXSA-N

Packaging

1 L quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 20 psig for 1 L. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

We are in the process of phasing-out the CGA 110/170 valves in favor of CGA 110/180 valves. Some stock of cylinders with CGA 110/170 valves may still exist. Please use adapters Z147095-1EA (brass) or Z147125-1EA (stainless steel) to utilize with a CGA 180 regulator.

Recommended products

Stainless steel control valves Z146951 or Z187224
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-306.4 °F - closed cup

Flash Point(C)

-188 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service