Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

486558

Sigma-Aldrich

Dimethyl-d6-amine

99 atom % D

Synonym(s):

Bis(trideuteriomethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2NH
CAS Number:
Molecular Weight:
51.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

vapor density

1.55 (15 °C, vs air)

vapor pressure

1277 mmHg ( 20 °C)

isotopic purity

99 atom % D

Assay

99% (CP)

autoignition temp.

753 °F

technique(s)

protein expression: suitable

bp

7 °C (lit.)

mp

−93 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])NC([2H])([2H])[2H]

InChI

1S/C2H7N/c1-3-2/h3H,1-2H3/i1D3,2D3

InChI key

ROSDSFDQCJNGOL-WFGJKAKNSA-N

Packaging

5 g quantity is packaged in a 450 mL carbon steel lecture bottle with stainless steel CGA 110/180 valve. Nominal gas pressure at 21ºC is 11 psig for 5 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z148504 and Z148512 and a "T"-purge valve Z151661 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Gas 1 - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

19.9 °F - closed cup

Flash Point(C)

-6.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service