Skip to Content
MilliporeSigma
All Photos(1)

Documents

617083

Sigma-Aldrich

Ethanol-1,1,2,2-d4-amine

98 atom % D

Synonym(s):

2-Amino(ethanol-1,1,2,2-d4)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCD2CD2NH2
Molecular Weight:
65.11
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

bp

170 °C (lit.)

mp

11 °C (lit.)

density

1.080 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]C([2H])(N)C([2H])([2H])O

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2/i1D2,2D2

InChI key

HZAXFHJVJLSVMW-LNLMKGTHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhiro Horibata et al.
Journal of lipid research, 61(8), 1221-1231 (2020-06-25)
The final step of the CDP-ethanolamine pathway is catalyzed by ethanolamine phosphotransferase 1 (EPT1) and choline/EPT1 (CEPT1). These enzymes are likely involved in the transfer of ethanolamine phosphate from CDP-ethanolamine to lipid acceptors such as 1,2-diacylglycerol (DAG) for PE production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service