Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

473669

Sigma-Aldrich

(Aminomethyl)polystyrene

200-400 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 2 % cross-linked

Synonym(s):

AM-polystyrene, Benzylamine polymer-bound, Poly(styrene-co-divinylbenzene), aminomethylated, Poly(styrene-co-vinylbenzylamine-co-divinylbenzene, Polystyrene crosslinked with divinylbenzene, aminomethylated

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Quality Level

crosslinking

2 % cross-linked

reaction suitability

reaction type: Boc solid-phase peptide synthesis

extent of labeling

1.0-2.0 mmol/g loading

particle size

200-400 mesh

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Scavenges acids, acid chlorides, anhydrides, aldehydes and other electrophiles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen J. et al.
Tetrahedron Letters, 40, 9195-9195 (1999)
Flynn, D.L.
Journal of the American Chemical Society, 119, 4874-4874 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service