Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

472107

Sigma-Aldrich

Tris(2-aminoethyl)amine, polymer-bound

extent of labeling: 3.5-5.0 mmol/g N loading, 1 % cross-linked with divinylbenzene

Synonym(s):

N-{2-[Bis(2-aminoethyl)amino]ethyl}aminomethyl–polystyrene, high loading

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

solid

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

3.5-5.0 mmol/g N loading

functional group

amine

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Creswell, M.W., et al.
Tetrahedron, 54, 3983-3983 (1998)
R.J. Booth et al.
Journal of the American Chemical Society, 119, 4882-4882 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service