Skip to Content
MilliporeSigma

8.55020

Aminomethylated polystyrene HL (100-200 mesh)

Novabiochem®

Synonym(s):

AM resin HL (100-200 mesh)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.22
UNSPSC Code:
13111023
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

General description

Base amino-functionalized polystyrene resin. Can be loaded with linkers bearing carboxylic acid functionalities.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Swelling Volume (in DMF): lot specific reslut
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.50 - 1.50 mmol/g
TNBS-Test (positive): passes test
TNBS-Test (after loading with Fmoc-Leu-OH (negative)): passes test
The polymer matrix is copoly (styrene -1% DVB), 100 -200 mesh.

Other Notes

Replaces: 01-64-0143

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library