Skip to Content
MilliporeSigma
All Photos(2)

Documents

459712

Sigma-Aldrich

Copper(II) 2,3,9,10,16,17,23,24-octakis(octyloxy)-29H,31H-phthalocyanine

Dye content 95 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C96H144CuN8O8
CAS Number:
Molecular Weight:
1601.77
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 95%

mp

>300 °C (lit.)

λmax

677 nm

SMILES string

CCCCCCCCOc1cc2c3nc(nc4n5[Cu]n6c(n3)c7cc(OCCCCCCCC)c(OCCCCCCCC)cc7c6nc8nc(nc5c9cc(OCCCCCCCC)c(OCCCCCCCC)cc49)c%10cc(OCCCCCCCC)c(OCCCCCCCC)cc8%10)c2cc1OCCCCCCCC

InChI

1S/C96H144N8O8.Cu/c1-9-17-25-33-41-49-57-105-81-65-73-74(66-82(81)106-58-50-42-34-26-18-10-2)90-97-89(73)101-91-75-67-83(107-59-51-43-35-27-19-11-3)84(108-60-52-44-36-28-20-12-4)68-76(75)93(98-91)103-95-79-71-87(111-63-55-47-39-31-23-15-7)88(112-64-56-48-40-32-24-16-8)72-80(79)96(100-95)104-94-78-70-86(110-62-54-46-38-30-22-14-6)85(69-77(78)92(99-94)102-90)109-61-53-45-37-29-21-13-5;/h65-72H,9-64H2,1-8H3;/q-2;+2

InChI key

GIAVAZMIHRPGQC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service