Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

274909

Sigma-Aldrich

Nickel(II) phthalocyanine-tetrasulfonic acid tetrasodium salt

Synonym(s):

Phthalocyanine-tetrasulfonic acid Nickel(II) complex tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H12N8Na4NiO12S4
CAS Number:
Molecular Weight:
979.40
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

reaction suitability

reagent type: catalyst
core: nickel

λmax

657 nm

SMILES string

[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1ccc2c3nc(nc4n5[Ni]n6c(nc7nc(nc5c8cc(ccc48)S([O-])(=O)=O)c9cc(ccc79)S([O-])(=O)=O)c%10ccc(cc%10c6n3)S([O-])(=O)=O)c2c1

InChI

1S/C32H16N8O12S4.4Na.Ni/c41-53(42,43)13-1-5-17-21(9-13)29-34-25(17)33-26-18-6-2-15(55(47,48)49)11-23(18)31(35-26)40-32-24-12-16(56(50,51)52)4-8-20(24)28(39-32)38-30-22-10-14(54(44,45)46)3-7-19(22)27(36-29)37-30;;;;;/h1-12H,(H4-2,33,34,35,36,37,38,39,40,41

InChI key

UAMQBHBCNOEWGQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service