Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

452521

Sigma-Aldrich

Iron(III) phthalocyanine-4,4′,4′′,4′′′-tetrasulfonic acid,compound with oxygen monosodium salt hydrate

Dye content 80 %

Synonym(s):

Iron Phthalocyanine Complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H15FeN8O14S4Na · xH2O
Molecular Weight:
942.60 (anhydrous basis)
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 80%

mp

>300 °C (lit.)

λmax

635 nm

SMILES string

O.[Na+].O=O.OS(=O)(=O)c1ccc2c3nc(nc4n5[Fe-]n6c(nc7nc(nc5c8cc(ccc48)S([O-])(=O)=O)c9ccc(cc79)S(O)(=O)=O)c%10ccc(cc%10c6n3)S(O)(=O)=O)c2c1

InChI

1S/C32H16N8O12S4.Fe.Na.O2.H2O/c41-53(42,43)13-1-5-17-21(9-13)29-33-25(17)37-30-22-10-14(54(44,45)46)2-6-18(22)27(34-30)39-32-24-12-16(56(50,51)52)4-8-20(24)28(36-32)40-31-23-11-15(55(47,48)49)3-7-19(23)26(35-31)38-29;;;1-2;/h1-12H,(H4-2,33,34,35,36,37,38,39,40,41,42,43,44,45,46,47,48,49,50,51,52);;;;1H2/q-2;2*+1;;/p-1

InChI key

LGPKILMWHLVKJQ-UHFFFAOYSA-M

Application

  • A homogeneous photoelectrochemical hydrogen sulfide sensor based on the electronic transfer mediated by tetrasulfophthalocyanine.: This study by Wang et al. (2020) explores the use of Iron(III) phthalocyanine tetrasulfonic acid sodium salt as a mediator in a photoelectrochemical hydrogen sulfide sensor. The sensor demonstrated high sensitivity and stability, highlighting its potential application in environmental monitoring and industrial processes (Wang et al., 2020).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service