Skip to Content
MilliporeSigma
All Photos(1)

Documents

415286

Sigma-Aldrich

Copper(II) 5,9,14,18,23,27,32,36-octabutoxy-2,3-naphthalocyanine

Dye content 90 %

Synonym(s):

5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine copper(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H88CuN8O8
CAS Number:
Molecular Weight:
1353.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, 90%

λmax

853 nm

SMILES string

CCCCOc1c2ccccc2c(OCCCC)c3c4nc(nc5n6[Cu]n7c(n4)c8c(OCCCC)c9ccccc9c(OCCCC)c8c7nc%10nc(nc6c%11c(OCCCC)c%12ccccc%12c(OCCCC)c5%11)c%13c(OCCCC)c%14ccccc%14c(OCCCC)c%10%13)c13

InChI

1S/C80H88N8O8.Cu/c1-9-17-41-89-65-49-33-25-26-34-50(49)66(90-42-18-10-2)58-57(65)73-81-74(58)86-76-61-62(70(94-46-22-14-6)54-38-30-29-37-53(54)69(61)93-45-21-13-5)78(83-76)88-80-64-63(71(95-47-23-15-7)55-39-31-32-40-56(55)72(64)96-48-24-16-8)79(84-80)87-77-60-59(75(82-77)85-73)67(91-43-19-11-3)51-35-27-28-36-52(51)68(60)92-44-20-12-4;/h25-40H,9-24,41-48H2,1-8H3;/q-2;+2

InChI key

JJIWQZDKAOMTKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mattias N D Svensson et al.
Science advances, 6(26), eaba4353-eaba4353 (2020-07-09)
Fibroblast-like synoviocytes (FLS) are joint-lining cells that promote rheumatoid arthritis (RA) pathology. Current disease-modifying antirheumatic agents (DMARDs) operate through systemic immunosuppression. FLS-targeted approaches could potentially be combined with DMARDs to improve control of RA without increasing immunosuppression. Here, we assessed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service