Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

407941

Sigma-Aldrich

2,2′-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol]

99%

Synonym(s):

2,2′-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-benzotriazol-2-ylphenol], 2,2′-Methylenebis[6-(2H -benzotriazol-2-yl)-4-tert -octylphenol], 2,2′-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H50N6O2
CAS Number:
Molecular Weight:
658.87
Beilstein:
11326835
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

mp

197-199 °C (lit.)

SMILES string

CC(C)(C)CC(C)(C)c1cc(Cc2cc(cc(c2O)-n3nc4ccccc4n3)C(C)(C)CC(C)(C)C)c(O)c(c1)-n5nc6ccccc6n5

InChI

1S/C41H50N6O2/c1-38(2,3)24-40(7,8)28-20-26(36(48)34(22-28)46-42-30-15-11-12-16-31(30)43-46)19-27-21-29(41(9,10)25-39(4,5)6)23-35(37(27)49)47-44-32-17-13-14-18-33(32)45-47/h11-18,20-23,48-49H,19,24-25H2,1-10H3

InChI key

FQUNFJULCYSSOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2′-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol] is a benzotriazole based additive that is used as an ultraviolet absorber (UVA). It reduces degradation of the polymeric material by absorbing the UV radiation and converting it to thermal heat.

Application

It is majorly used as a UV filter in sunscreens and in photo-luminescent dyes for organic electronic based applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple extraction and HPLC determination of UV-A and UV-B filters in sunscreen products.
De Orsi D, et al.
Chromatographia, 64(9-10), 509-515 (2006)
Effect of polymer matrix on photo-stability of photo-luminescent dyes in multi-layer polymeric structures.
Baumberg I, et al.
Polymer Degradation and Stability, 73(3), 403-410 (2001)
Analysis of UV absorbers and stabilizers in polypropylene by liquid chromatography/atmospheric pressure chemical ionization-mass spectrometry.
Choi S and Jang J
Polymer Testing, 30(6), 673-677 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service