Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

30184

Supelco

Bisoctrizole

analytical standard

Synonym(s):

2,2′-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol], Tinosorb M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H50N6O2
CAS Number:
Molecular Weight:
658.87
Beilstein:
11326835
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

195-199 °C
197-199 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(C)(C)CC(C)(C)c1cc(Cc2cc(cc(c2O)-n3nc4ccccc4n3)C(C)(C)CC(C)(C)C)c(O)c(c1)-n5nc6ccccc6n5

InChI

1S/C41H50N6O2/c1-38(2,3)24-40(7,8)28-20-26(36(48)34(22-28)46-42-30-15-11-12-16-31(30)43-46)19-27-21-29(41(9,10)25-39(4,5)6)23-35(37(27)49)47-44-32-17-13-14-18-33(32)45-47/h11-18,20-23,48-49H,19,24-25H2,1-10H3

InChI key

FQUNFJULCYSSOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bisoctrizole (UV 360) belongs to the class of benzotriazole UV stabilizers, commercially used in sunscreen formulations. Its mode of action involves a triple effect including UV absorption, light scattering and light reflection. The highly photostable nature of UV 360, makes it feasible to stabilize other organic filters such as avobenzene.

Application

Bisoctrizole may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Packaging materials using high performance liquid chromatography (HPLC) with a diode array detector (DAD).
  • Seawater samples using ultra-high-performance liquid chromatography and tandem mass spectrometry detection (UHPLC-MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction and determination methodologies for benzotriazole UV stabilizers in personal-care products in environmental and biological samples
Montesdeoca-Esponda S, et al.
TrAC, Trends in Analytical Chemistry, 51, 23-32 (2013)
Determination of UV filters in packaging by focused ultrasonic solid?liquid extraction and liquid chromatography
Moreta C, et al.
Journal of Chromatography A, 1218(21), 3392-3399 (2011)
Rapid monitoring of residual UV-stabilizers in seawater samples from beaches using fabric phase sorptive extraction and UHPLC-MS/MS
Garcia-Guerra BR, et al.
Chemosphere, 164, 201-207 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service