Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

795682

Sigma-Aldrich

(4R,4′R,5S,5′S)-2,2′-(1-Methylethylidene

95%

Synonym(s):

2,2-Bis[2-(4R,5S-diphenyl-1,3-oxazolinyl)]propane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H30N2O2
CAS Number:
Molecular Weight:
486.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

164-169 °C

SMILES string

CC(C1=N[C@H](C2=CC=CC=C2)[C@H](C3=CC=CC=C3)O1)(C)C4=N[C@H](C5=CC=CC=C5)[C@H](C6=CC=CC=C6)O4

InChI

1S/C33H30N2O2/c1-33(2,31-34-27(23-15-7-3-8-16-23)29(36-31)25-19-11-5-12-20-25)32-35-28(24-17-9-4-10-18-24)30(37-32)26-21-13-6-14-22-26/h3-22,27-30H,1-2H3/t27-,28-,29+,30+/m1/s1

InChI key

ZWWGNCSTEMMQOQ-XAZDILKDSA-N

Application

(4R,4′R,5S,5′S)-2,2′-(1-Methylethylidene) or (4S,5R)-Bis-Phbox) can be used as a ligand:
  • To prepare selective exo-catalysts for enantioselective 1,3-dipolar cycloaddition reactions.
  • In the asymmetric aminooxygenation of alkenes in the presence of tetramethylaminopyridyl radical (TEMPO) as an oxidant and copper(II) triflate as a catalyst.
  • In asymmetric aminofluorination of olefins using an iron catalyst.

This chiral Box ligand was most recently shown to mediate an asymmetric aminofluorination of olefins utlizing Xtalfuor-E (719439) and TREAT-HF (344648). The resulting cyclic carbamates can be readily converted into their concomitant beta-fluoro amino acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper catalyzed enantioselective intramolecular aminooxygenation of alkenes
Fuller PH, et al.
Journal of the American Chemical Society, 130(52), 17638-17639 (2008)
Iron (II)-catalyzed intramolecular olefin aminofluorination
Lu Deng-Fu, et al.
Organic Letters, 16(11), 2912-2915 (2014)
In Search of exo-Selective Catalysts for Enantioselective 1, 3-Dipolar Cycloaddition between Acryloyloxazolidinone and Diphenylnitrone
Desimoni G, et al.
European Journal of Organic Chemistry, 2005(6), 1020-1027 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service