Skip to Content
MilliporeSigma

413151

2-Hydroxy-4-(octyloxy)benzophenone

98%

Synonym(s):

2-Benzoyl-5-octyloxyphenol, 4-(Octyloxy)-2-hydroxybenzophenone, [2-Hydroxy-4-(octyloxy)phenyl]phenylmethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)7OC6H3(OH)COC6H5
CAS Number:
Molecular Weight:
326.43
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12162002
EC Number:
217-421-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

98%

mp

47-49 °C (lit.)

SMILES string

CCCCCCCCOc1ccc(c(O)c1)C(=O)c2ccccc2

InChI

1S/C21H26O3/c1-2-3-4-5-6-10-15-24-18-13-14-19(20(22)16-18)21(23)17-11-8-7-9-12-17/h7-9,11-14,16,22H,2-6,10,15H2,1H3

InChI key

QUAMTGJKVDWJEQ-UHFFFAOYSA-N

General description

2-Hydroxy-4-(octyloxy)benzophenone (HOBP) is a UV based absorber that has 2-hydroxybenzophenone as a functional group. It can be used to enhance the light fastness of polymers using UV absorbers with sulfur and phosphorus compounds.

Application

HOBPs are UV absorbers for insulating plastics, which can be incorporated with active layers to increase the stabilization of organic solar cells (OSCs). It may be incorporated in parylene C films to enhance the light stability, which can be potentially used as a moisture transfer membrane for electronic boards and photovoltaics.


Still not finding the right product?

Explore all of our products under 2-Hydroxy-4-(octyloxy)benzophenone


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1B

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

215.6 °F - closed cup

flash_point_c

102 °C - closed cup

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library