Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

395048

Sigma-Aldrich

Methylamine solution

greener alternative

2.0 M in methanol

Synonym(s):

Monomethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH2
CAS Number:
Molecular Weight:
31.06
Beilstein:
741851
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

10.86 psi ( 55 °C)
2.07 psi ( 20 °C)

Quality Level

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

2.0 M in methanol

bp

40 °C

density

0.785 g/mL at 25 °C

functional group

amine

greener alternative category

SMILES string

CN

InChI

1S/CH5N/c1-2/h2H2,1H3

InChI key

BAVYZALUXZFZLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Methylamine solution is an aqueous solution of Methylamine. It is used to synthesize amino-terminated monolayers of phthalimide on silicon surfaces.

Application

Methylamine solution 2.0 M in methanol can be used as a precursor that facilitates the synthesis of perovskite based materials. It can also be used as a deactivating agent to study the catalytic performance of acidic macroporous ion exchange resins. It can be annealed to reduce impurity in the grain boundaries, improve the stability and efficiency of the perovskite solar cells.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of amino-terminated monolayers via hydrolysis of phthalimide anchored to Si (1 1 1)
Ara, Masato and Tsuji
Surface Science, 601, 5098-5510 (2007)
Deactivation of ion exchange catalysts by acetonitrile and methylamine
Gonzalez R, et al.
Topics in Catalysis, 54(16-18), 1054-1054 (2011)
Post-annealing of MAPbI 3 perovskite films with methylamine for efficient perovskite solar cells
Jiang Y, et al.
Materials Horizons, 3(6), 548-555 (2016)
Xin Yu Chin et al.
Nature communications, 6, 7383-7383 (2015-06-26)
Despite the widespread use of solution-processable hybrid organic-inorganic perovskites in photovoltaic and light-emitting applications, determination of their intrinsic charge transport parameters has been elusive due to the variability of film preparation and history-dependent device performance. Here we show that screening
Jeff O'Sullivan et al.
Neurotoxicology, 25(1-2), 303-315 (2003-12-31)
The semicarbazide-sensitive amine oxidases (SSAO) (EC 1.4.3.6) were believed to be detoxifying enzymes, primarily involved in the oxidative deamination of endogenous amines, such as methylamine and aminoacetone, together with some xenobiotic amines. However, it appears that the reaction products may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service