Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345830-M

Sigma-Aldrich

Galectin-3, Human, Recombinant, E. coli

Synonym(s):

CBP30, CBP35, εBP, L-29, L-34, Mac-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

≤1.0 EU/μg Endotoxin (EU/μg galectin-3)

storage temp.

−20°C

General description

Recombinant, human galectin-3 expressed in and purified from E. coli. Galectin-3 is a member of the β-galactoside-binding animal lectins. It has been implicated to be involved in various immunological processes, pre-mRNA splicing, cell adhesion, and tumor invasion. Levels of galectin-3 have been reported to be elevated in several metastatic cancers.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from diH₂O.

Reconstitution

Following reconstitution refrigerate (4°C). Stock solutions containing lactose are stable for up to 6 months at 4°C. Following dialysis and removal of lactose, stock solutions are stable for up to 4 days at 4°C.
Reconstitute in sterile PBS. For use in carbohydrate binding studies, lactose should be removed by dialysis.

Other Notes

Legendre, H., et al. 2003. Mod. Pathol.16, 491.
Maeda, M., et al. 2003. J. Biol. Chem.278, 18938.
Stock, M., et al. 2003. J. Biol. Chem.278, 17360.
Hirabayashi, J., et al. 2002. Biochim. Biophys. Acta1572, 232.
Iurisci, I., et al. 2000. Clin. Cancer Res.6, 1389.
Oda, Y., et al. 1991. Gene99, 279.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service