Skip to Content
MilliporeSigma
All Photos(2)

Documents

499145

Sigma-Aldrich

Ammonia solution

7 N in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH3
CAS Number:
Molecular Weight:
17.03
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

form

liquid

concentration

6.5-7.5 M (by NaOH, titration)
7 N in methanol

impurities

≤0.1% water (Karl Fischer)

density

0.779 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

N

InChI

1S/H3N/h1H3

InChI key

QGZKDVFQNNGYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonia solution is commonly used as a pH-adjusting or buffering agent in a variety of liquid formulations. It also acts as a base in some chemical reactions.

Application

Ammonia solution (7N methanolic ammonia) can be used in the synthesis of:      
  • Hydrogen halide salts of primary amines from corresponding halides under microwave irradiation.      
  • N-alkylbis(3-aminopropyl)amines by Raney nickel catalyzed hydrogenation of N-alkylbis(cyanoethyl)amines.
  • 2-quinoxalinecarboxamide by ammonolysis of 2-methoxycarbonylquinoxaline.

Ammonia solution (7N methanolic ammonia) may be used in the preparation of trioxolanes. It may also be used in the preparation of hydrogen halide salts (HX) of primary amines under microwave irradiation.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weak base dispiro-1,2,4-trioxolanes: potent antimalarial ozonides.
Tang Y, et al.
Bioorganic & Medicinal Chemistry Letters, 17(5), 1260-1265 (2007)
Microwave-assisted synthesis of primary amine HX salts from halides and 7M ammonia in methanol.
Saulnier MG, et al.
Tetrahedron Letters, 45(2), 397-399 (2004)
Eagleson M.
Concise Encyclopedia Chemistry, 65-66 (1994)
Ledgard J.
The Preparatory Manual of Explosives., 41-41 (2007)
Patrick M Erwin et al.
The ISME journal, 8(3), 575-588 (2013-10-25)
Ascidians are ecologically important components of marine ecosystems yet the ascidian microbiota remains largely unexplored beyond a few model species. We used 16S rRNA gene tag pyrosequencing to provide a comprehensive characterization of microbial symbionts in the tunic of 42

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service