Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ARK2209

SAFC

Methylamine

33% w/w in Ethanol Denatured with Toluene(1%)

Pharma Manufacturing

Synonym(s):

Monomethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH2
CAS Number:
Molecular Weight:
31.06
Beilstein:
741851
MDL number:
UNSPSC Code:
46182005

vapor density

1.08 (20 °C, vs air)

Quality Level

vapor pressure

27 psi ( 20 °C)

autoignition temp.

806 °F

expl. lim.

4.9-20.8 %

bp

-6.3 °C (lit.)

mp

-93 °C (lit.)

density

0.70 g/mL at 20 °C (lit.)

SMILES string

CN

InChI

1S/CH5N/c1-2/h2H2,1H3

InChI key

BAVYZALUXZFZLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-9.0 °F - closed cup

Flash Point(C)

-22.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service