Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

337781

Sigma-Aldrich

Tetradecylmagnesium chloride solution

1.0 M in THF

Synonym(s):

Chlorotetradecylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13MgCl
CAS Number:
Molecular Weight:
257.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

density

0.923 g/mL at 25 °C

SMILES string

CCCCCCCCCCCCCC[Mg]Cl

InChI

1S/C14H29.ClH.Mg/c1-3-5-7-9-11-13-14-12-10-8-6-4-2;;/h1,3-14H2,2H3;1H;/q;;+1/p-1

InChI key

SKZDCNLZNNZOFG-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service