Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

630551

Sigma-Aldrich

(2-Ethylhexyl)magnesium bromide solution

1.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17BrMg
CAS Number:
Molecular Weight:
217.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

Quality Level

concentration

1.0 M in diethyl ether

density

0.863 g/mL at 25 °C

SMILES string

CCCCC(CC)C[Mg]Br

InChI

1S/C8H17.BrH.Mg/c1-4-6-7-8(3)5-2;;/h8H,3-7H2,1-2H3;1H;/q;;+1/p-1

InChI key

NDCJRLBSQQDNOO-UHFFFAOYSA-M

Application

(2-Ethylhexyl)magnesium bromide (Grignard reagent) can be used in the synthesis of:       
  • 3-ethyl-N,N-dimethyl-1-(pyridin-2-yl)heptan-1-amine, a pyridyl amine ligand, which is used in the polymerization of olefins.   
  •  Diphenylbis(2-ethylhexyl) germane from diphenylgermanium dichloride by Grignard addition reaction.       
  • Dichloro (dodecyl) 2-ethylhexylsilane, a key intermediate that is used in the preparation of silafluorene-based copolymers.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-13.0 °F - closed cup

Flash Point(C)

-25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A practical synthesis of N, N-dimethyl-(6-arylpyrid-2-yl) alkylamines
Faler, Catherine A
Tetrahedron Letters, 51(42), 5621-5623 (2010)
A low band gap co-polymer of dithienogermole and 2, 1, 3-benzothiadiazole by Suzuki polycondensation and its application in transistor and photovoltaic cells
Fei Z, et al.
Journal of Materials Chemistry, 21(40), 16257-16263 (2011)
Chinna D Bathula et al.
Journal of nanoscience and nanotechnology, 14(8), 6002-6007 (2015-05-06)
A series of silafluorene-based copolymers, poly[9-(2-ethylhexyl)-9-dodecyl-silafluorene-2,7-diyl-alt-5,5-(4',7'-di-2-thienyl-2',1',3'-benzothiadiazole)] (P1), poly[9-(2-ethylhexyl)-9-dodecyl-silafluorene-2,7-diyl-alt-2,5-bis-(thiophene-2-yl)thiazolo [5,4-d]thiazole] (P2), and poly[9-(2-ethylhexyl)-9-dodecyl-silafluorene-2,7-diyl-alt-5,5-(5',8'-di-2-thienyl-2,3-bis(4-octyloxyl)phenyl)quinoxaline] (P3), were synthesized and used as donor materials in polymer solar cells (PSCs). The optical, electrochemical, and photovoltaic properties of the copolymers were investigated. The results indicate that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service