Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

424056

Sigma-Aldrich

Pentadecylmagnesium bromide solution

0.5 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14MgBr
CAS Number:
Molecular Weight:
315.62
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in diethyl ether

bp

34.6 °C

density

0.793 g/mL at 25 °C

SMILES string

CCCCCCCCCCCCCCC[Mg]Br

InChI

1S/C15H31.BrH.Mg/c1-3-5-7-9-11-13-15-14-12-10-8-6-4-2;;/h1,3-15H2,2H3;1H;/q;;+1/p-1

InChI key

NNIBYWYQYUJVFG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinlin Li et al.
Molecular imaging, 14, 516-525 (2015-10-16)
The goal of this study was to establish a quantitative method for measuring fatty acid (FA) metabolism with partial volume (PV) and spill-over (SP) corrections using dynamic [(11)C]palmitate positron emission tomographic (PET) images of mouse heart in vivo. Twenty-minute dynamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service