Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

317306

Sigma-Aldrich

3,3′-Iminodipropionitrile

technical grade, 90%

Synonym(s):

Bis(2-cyanoethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2CN)2
CAS Number:
Molecular Weight:
123.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.47 (lit.)

bp

205 °C/25 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

N#CCCNCCC#N

InChI

1S/C6H9N3/c7-3-1-5-9-6-2-4-8/h9H,1-2,5-6H2

InChI key

SBAJRGRUGUQKAF-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serkan Demirci et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 99, 144-149 (2012-10-16)
The chemical conversion of the top surface of 3-glycidoxypropyldimethylethoxysilane (GPDMES) on hyroxylated silicon surface has been studied as a function of reaction time. A multiple-step procedure was applied in this study. At first, GPDMES molecules were self-assembled on the hydroxylated
M Tariq et al.
Behavioural pharmacology, 15(8), 585-588 (2004-12-04)
This study reports the effects of the antioxidant sodium benzoate (SB) on iminodipropionitrile (IDPN)-induced excitation with choreiform and circling (ECC) syndrome in adult female Wistar rats. Rats in four different groups (n=8) received i.p. injections of SB (0, 50, 100
Isabel Ding et al.
Biochimica et biophysica acta. Molecular cell research, 1867(9), 118739-118739 (2020-05-12)
The formation of extensions in cell migration requires tightly coordinated reorganization of all three cytoskeletal polymers but the mechanisms by which intermediate filament networks interact with actin to generate extensions are not well-defined. We examined interactions of the actin binding
Haseeb Ahmad Khan et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 55(2-3), 181-186 (2003-11-19)
A direct association between aging and drug-induced dyskinesia has been reported by several investigators. Iminiodipropionitrile (IDPN), a prototype nitrile compound produces a motor syndrome in rodents, which resembles neuroleptic drug induced dyskinesia. In this investigation attempt has been made to
Pengcheng Zhu et al.
Experimental and therapeutic medicine, 21(1), 51-51 (2020-12-05)
Quantitative evaluation of tic disorders (TDs) is challenging as there are few objective indicators that can be used for the assessment of treatment outcomes. 18F-Fluorodeoxyglucose (FDG) is a radioactive tracer that is able to cross the blood-brain barrier and can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service