Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239399

Sigma-Aldrich

1,2-Bis(3-aminopropylamino)ethane

technical grade, 94%

Synonym(s):

3,3′-Ethylenediiminodipropylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2CH2NHCH2CH2NHCH2CH2CH2NH2
CAS Number:
Molecular Weight:
174.29
Beilstein:
2203748
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

94%

refractive index

n20/D 1.491 (lit.)

bp

150-160 °C/5 mmHg (lit.)

density

0.952 g/mL at 25 °C (lit.)

SMILES string

NCCCNCCNCCCN

InChI

1S/C8H22N4/c9-3-1-5-11-7-8-12-6-2-4-10/h11-12H,1-10H2

InChI key

RXFCIXRFAJRBSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The condensation reaction of 1,2-bis(3-aminopropylamino)ethane [3,3′-ethylenediiminodipropylamine] with 4,6-dibenzofurandicarbaldehyde was studied.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New dioxadiaza-, trioxadiaza-and hexaaza-macrocycles containing dibenzofuran units.
Tetrahedron, 62(36), 8550-8558 (2006)
David R Goode et al.
Journal of medicinal chemistry, 51(8), 2346-2349 (2008-03-28)
It is recognized that high-throughput enzyme inhibition screens often return nonspecific inhibitors as "hits". Recently, high-throughput screens for enzyme activators have led to the identification of several compounds with novel and potent biological activity. Here, we show that enzyme activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service