263885
2,4,6-Trimethyl-m-phenylenediamine
96%
Synonym(s):
2,4,6-Trimethyl-1,3-phenylenediamine, 2,4-Diamino-1,3,5-trimethylbenzene, 2,4-Diaminomesitylene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
96%
form
solid
mp
89-91 °C (lit.)
SMILES string
Cc1cc(C)c(N)c(C)c1N
InChI
1S/C9H14N2/c1-5-4-6(2)9(11)7(3)8(5)10/h4H,10-11H2,1-3H3
InChI key
ZVDSMYGTJDFNHN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Membranes, 10(4) (2020-04-23)
The energy-intensive ethylene/ethane separation process is a key challenge to the petrochemical industry. HKUST-1, a metal-organic framework (MOF) which possesses high accessible surface area and porosity, is utilized in mixed-matrix membrane fabrication to investigate its potential for improving the performance
Membranes, 10(7) (2020-07-28)
Nanocrystalline UiO-66 and its derivatives (containing -NH2, -Br, -(OH)2) were developed via pre-synthetic functionalization and incorporated into a polyimide membrane to develop a mixed-matrix membrane (MMM) for CO2/N2 separation. Incorporation of the non-functionalized UiO-66 nanocrystals into the polyimide membrane successfully
ChemSusChem, 13(19), 5318-5328 (2020-07-31)
Partial substitution of the asymmetric 3,3',4,4'-biphenyl dianhydride monomer (aBPDA) into the backbone of a 6FDA-BPDA-DAM (6FDA=4,4'-hexafluoroisopropylidene, DAM=diaminomesitylene) diphthalic anhydride-based copolyimide based on symmetrical BPDA (sBPDA) was used to study membrane structure-processing-property relationships for gas separation. Properties of the polymer membrane
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service