Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

368148

Sigma-Aldrich

4,4′-(Hexafluoroisopropylidene)dianiline

98%

Synonym(s):

2,2-Bis(4-aminophenyl)hexafluoropropane, 4,4′-(Hexafluoroisopropylidene)dianiline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C(C6H4NH2)2
CAS Number:
Molecular Weight:
334.26
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

195-198 °C (lit.)

SMILES string

Nc1ccc(cc1)C(c2ccc(N)cc2)(C(F)(F)F)C(F)(F)F

InChI

1S/C15H12F6N2/c16-14(17,18)13(15(19,20)21,9-1-5-11(22)6-2-9)10-3-7-12(23)8-4-10/h1-8H,22-23H2

InChI key

BEKFRNOZJSYWKZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tae Soo Jo et al.
Polymers, 11(5) (2019-05-15)
A set of poly(pyridinium) salts containing various diamine moieties, as molecular connectors, and poly(ethyleneglycol)-4-nonylphenyl-3-sulfopropyl ether, thereafter referred to as "Macroion", as the hydrophilic counterion, were prepared by metathesis reaction from the respective precursory tosylated poly(pyridinium)s in methanol. The structure of

Articles

Fluorocarbon polymers exhibit increased thermal stability, hydrophobicity, lipophobicity, and chemical resistance compared to hydrocarbon analogs.

Fluorocarbon polymers exhibit increased thermal stability, hydrophobicity, lipophobicity, and chemical resistance compared to hydrocarbon analogs.

Fluorocarbon polymers exhibit increased thermal stability, hydrophobicity, lipophobicity, and chemical resistance compared to hydrocarbon analogs.

Fluorocarbon polymers exhibit increased thermal stability, hydrophobicity, lipophobicity, and chemical resistance compared to hydrocarbon analogs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service