Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

242691

Sigma-Aldrich

3-Iodoanisole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4OCH3
CAS Number:
Molecular Weight:
234.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.613 (lit.)

bp

244-245 °C (lit.)

density

1.965 g/mL at 25 °C (lit.)

functional group

iodo

SMILES string

COc1cccc(I)c1

InChI

1S/C7H7IO/c1-9-7-4-2-3-6(8)5-7/h2-5H,1H3

InChI key

RSHBAGGASAJQCH-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elaine Crosbie et al.
Dalton transactions (Cambridge, England : 2003), 41(6), 1832-1839 (2011-12-14)
Gaining a deeper understanding of the modus operandi of heterometallic lithium aluminate bases towards deprotonative metallation of substituted aromatic substrates, we have studied the reactions and their aftermath between our recently developed bis-amido base '(i)Bu(2)Al(μ-TMP)(2)Li'3 and 3-halogenated anisoles. Ortho-metallation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service