Skip to Content
MilliporeSigma
All Photos(1)

Documents

238139

Sigma-Aldrich

1-Iodonaphthalene

97%

Synonym(s):

1-Naphthyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7I
CAS Number:
Molecular Weight:
254.07
Beilstein:
1906415
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.701 (lit.)

bp

163-165 °C/15 mmHg (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

Ic1cccc2ccccc12

InChI

1S/C10H7I/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

NHPPIJMARIVBGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ultrafast relaxation of 1-iodonaphthalene has been studied by time-resolved femtosecond pump-probe mass spectrometry. 1-Iodonaphthalene undergoes Pd catalyzed cross-coupling reaction (Stille reaction) with 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine to afford 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raul Montero et al.
Physical chemistry chemical physics : PCCP, 12(28), 7988-7993 (2010-06-03)
The ultrafast relaxation of 1-iodonaphthalene, with particular attention to the dissociation channels, has been studied by time-resolved femtosecond pump-probe mass spectrometry following excitation at 267 and 317 nm. The measured transients for the parent ion and the isobaric fragments, iodine
Javier I Bardagí et al.
The Journal of organic chemistry, 73(12), 4491-4495 (2008-05-21)
From the commercial 6-chloro-2,4-dimethoxypyrimidine (1) and by a photostimulated reaction with Me(3)Sn(-) ions, 2,4-dimethoxy-6-(trimethylstannyl)pyrimidine (2) was obtained in 95% yield. By the cross-coupling reaction of 2 with 1-iodonaphthalene as electrophile catalyzed by Pd (Stille reaction), 2,4-dimethoxy-6-(naphthalen-1-yl)pyrimidine (9) was obtained in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service