Skip to Content
MilliporeSigma
All Photos(1)

Documents

252786

Sigma-Aldrich

2-Iodoanisole

98%

Synonym(s):

2-Iodophenyl methyl ether, 2-Methoxyiodobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4OCH3
CAS Number:
Molecular Weight:
234.03
Beilstein:
1860243
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.622 (lit.)

bp

125-126 °C/19 mmHg (lit.)

solubility

alcohol: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
water: insoluble(lit.)

density

1.799 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1I

InChI

1S/C7H7IO/c1-9-7-5-3-2-4-6(7)8/h2-5H,1H3

InChI key

DVQWNQBEUKXONL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Iodoanisole participates in palladium catalyzed enantioselective Heck arylation of 2,3-dihydrofuran in the presence of chiral ionic liquids containing L-prolinate and L-lactate anions and non-chiral quaternary ammonium cations.

Application

2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Morel et al.
Dalton transactions (Cambridge, England : 2003), 42(4), 1215-1222 (2012-11-09)
Chiral ionic liquids (CILs) containing L-prolinate and L-lactate anions and non-chiral quaternary ammonium cations were employed in the palladium catalyzed enantioselective Heck arylation of 2,3-dihydrofuran with aryl iodides (iodobenzene, 4-iodotoluene, 2-iodoanisole, 4-iodoanisole, 4-iodoacetophenone). In all the reactions 2-aryl-2,3-dihydrofuran (3) was
Dawei Yue et al.
The Journal of organic chemistry, 70(25), 10292-10296 (2005-12-06)
[reaction: see text] 2,3-Disubstituted benzo[b]furans are readily prepared under very mild reaction conditions by the palladium/copper-catalyzed cross-coupling of various o-iodoanisoles and terminal alkynes, followed by electrophilic cyclization with I2, PhSeCl, or p-O2NC6H4SCl. Aryl- and vinylic-substituted alkynes undergo electrophilic cyclization in
Naoyuki Yasaka et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(10), 1183-1188 (2018-10-12)
Aryl halides are a very important category of compounds that include many vital drugs and key industrial additives, such as clofibrate and bromobenzene, respectively. Due to their importance, our research group previously developed a novel fluorescence labeling approach for their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service