Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

176702

Sigma-Aldrich

Ammonium-d4 deuteroxide solution

25 wt. % in D2O, 99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ND4OD
CAS Number:
Molecular Weight:
40.08
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

concentration

25 wt. % in D2O

technique(s)

NMR: suitable
bio NMR: suitable

refractive index

n20/D 1.334

density

1.056 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]O[2H].[2H]N([2H])[2H]

InChI

1S/H3N.H2O/h1H3;1H2/i/hD5

InChI key

VHUUQVKOLVNVRT-NSPFYZSMSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service