Skip to Content
MilliporeSigma
All Photos(1)

Documents

369632

Sigma-Aldrich

Trifluoromethanesulfonic acid-d

98 atom % D

Synonym(s):

Deuterotrifluormethansulfonic acid, Triflic acid-D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3D
CAS Number:
Molecular Weight:
151.08
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

form

liquid

bp

162 °C (lit.)

density

1.708 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OS(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S/c2-1(3,4)8(5,6)7/h(H,5,6,7)/i/hD

InChI key

ITMCEJHCFYSIIV-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service