Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

157120

Sigma-Aldrich

4-tert-Butylbenzoyl chloride

98%

Synonym(s):

4-(1,1-Dimethylethyl)benzoyl chloride, 4-tert-Butylbenzoic acid chloride, p-tert-Butylbenzoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4COCl
CAS Number:
Molecular Weight:
196.67
Beilstein:
775793
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.536 (lit.)

bp

135 °C/20 mmHg (lit.)

density

1.007 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

CC(C)(C)c1ccc(cc1)C(Cl)=O

InChI

1S/C11H13ClO/c1-11(2,3)9-6-4-8(5-7-9)10(12)13/h4-7H,1-3H3

InChI key

WNLMYNASWOULQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

AlCl3 promoted Friedel-Crafts acylation of 4-tert-butylbenzoyl chloride with mesitylene was investigated.

Application

4-tert-Butylbenzoyl chloride was used in the synthesis of 9,9′-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties. It was also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuh-Yih Chien et al.
Chemical communications (Cambridge, England), (23)(23), 2874-2875 (2002-12-14)
Some 9,9'-spirobifluorene-bridged bipolar systems 1-3 containing 1,3,4-oxadiazole-conjugated oligoaryl and triarylamine moieties have been synthesized, in which 1 exhibits remarkable solvent-polarity dependent fluorescence properties due to a highly efficient photoinduced electron transfer reaction.
Statistical copolymers with side-chain hole and electron transport groups for single-layer electroluminescent device applications.
Jiang X, et al.
Chemistry of Materials, 12(9), 2542-2549 (2000)
Zhiliang Huang et al.
Organic & biomolecular chemistry, 11(11), 1810-1814 (2013-02-08)
AlCl(3) promoted Friedel-Crafts acylation between 4-tert-butylbenzoyl chloride and mesitylene was investigated. The donor-acceptor complex was observed as the major species. Kinetic investigation demonstrated that the reaction was first-order on the donor-acceptor complex and zero-order on ArH, suggesting that the donor-acceptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service