Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

122254

Sigma-Aldrich

m-Toluoyl chloride

99%

Synonym(s):

3-Methylbenzoyl chloride, m-Toluic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COCl
CAS Number:
Molecular Weight:
154.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.5485 (lit.)

bp

86 °C/5 mmHg (lit.)

density

1.173 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

Cc1cccc(c1)C(Cl)=O

InChI

1S/C8H7ClO/c1-6-3-2-4-7(5-6)8(9)10/h2-5H,1H3

InChI key

YHOYYHYBFSYOSQ-UHFFFAOYSA-N

General description

m-Toluoyl chloride is an efficient derivatization reagent for amines.

Application

m-Toluoyl chloride has been used to derivatize the C1-C4 aliphatic amines for their determination in air by HPLC. It has been used in determination of atmospheric ammonia by denuder-sampling and HPLC-UV detection.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved HPLC determination of aliphatic amines in air by diffusion and derivatization techniques.
Possanzini M and Di Palo V.
Chromatographia, 29(3-4), 151-154 (1990)
Determination of atmospheric ammonia as m-toluamide by denuder sampling and HPLC-UV detection.
Possanzini M and Di Palo V.
Chromatographia, 28(1-2), 27-30 (1989)
Ataf Ali Altaf et al.
Dalton transactions (Cambridge, England : 2003), 47(34), 11769-11781 (2018-08-18)
Twenty-three compounds in two series of ferrocene-based anilides, with the general formula C5H5-Fe-C5H4-C6H4-NH-CO-C6H4-R (where R = H, F, Cl, CH3 and OCH3), have been successfully synthesized. The compounds were characterized by elemental analysis and FTIR, 1H NMR and 13C NMR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service